Learn More
Sino Biological Human ALCAM / CD166 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10045H08H200
A DNA sequence encoding the extracellular domain (Met 1-Ala 526) of human ALCAM (NP_001618.2) was expressed, with the C-terminal polyhistidine tag.
Catalog No. 50-161-3870