Learn More
Sino Biological CD166 / ALCAM Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10045MM03200
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD166 / ALCAM (rh CD166 / ALCAM; Catalog#10045-H08H; NP_001618.2; Met1-Ala526). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-161-1256