Learn More
Sino Biological CD166 / ALCAM Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10045RP02100
Produced in rabbits immunized with purified, recombinant Human CD166 / ALCAM (rh CD166 / ALCAM; Catalog#10045-H08H; NP_001618.2; Met 1-Ala 526). CD166 / ALCAM specific IgG was purified by Human CD166 / ALCAM affinity chromatography.
Catalog No. 50-161-1270