Learn More
Sino Biological CCR2 Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 100450MM02200
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with a synthetic peptide corresponding to the center region of the Human CCR2. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-161-1223