Learn More
Sino Biological CCR2 Antibody (APC), Mouse MAb DFS Item
Manufacturer:  Sino Biological 100450MM02A100
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with a synthetic peptide corresponding to the center region of the Human CCR2, and it was conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-161-1225