Learn More
Sino Biological CCR2/CD192 Antibody (APC), Mouse MAb 25Tests
Supplier:  Sino Biological 100450MM02A25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with a synthetic peptide corresponding to the center region of the Human CCR2/CD192, and it was conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-161-1226