Learn More
Sino Biological CCR2 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100450T3450
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human CCR2, and purified by antigen affinity chromatography.
Catalog No. 50-161-1236