Learn More
Sino Biological CCR3 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100451T38100
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human CCR3, and purified by antigen affinity chromatography.
Catalog No. 50-161-1240