Learn More
Sino Biological LY75 / DEC-205 / CD205 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100454T0850
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human LY75 / DEC-205 / CD205, and purified by antigen affinity chromatography.
Catalog No. 50-161-1248