Learn More
Sino Biological cd215 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100455T3250
Produced in rabbits immunized with A synthetic peptide corresponding to the center region of the human cd215, and purified by antigen affinity chromatography.
Catalog No. 50-161-1251