Learn More
Sino Biological CD61 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100458T0850
Produced in rabbits immunized with A synthetic peptide corresponding to the N-terminus of the human CD61, and purified by antigen affinity chromatography.
Catalog No. 50-161-1254