Learn More
Sino Biological SPARCL1 / SPARC-like 1 Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10046MM051
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SPARCL1 / SPARC-like 1 (rh SPARCL1 / SPARC-like 1; Catalog#10046-H08H; NP_004675.3; Met1-Phe664). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-161-1295