Learn More
Sino Biological SPARCL1 / MAST9 Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10046MM0650
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SPARCL1 (rhSPARCL1; Catalog#10046-H08H; NP_004675.3; Met 1-Phe 664). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-161-1303