Learn More
Sino Biological SPARCL1 / MAST9 Antibody, Rabbit MAb DFS Item
Manufacturer:  Sino Biological 10046R20450
This antibody was obtained from a rabbit immunized with purified, recombinant Human SPARCL1 (rhSPARCL1; Catalog#10046-H08H; NP_004675.3; Met 1-Phe 664).
Catalog No. 50-161-1309