Learn More
Sino Biological SPARCL1 Antibody (PE), Rabbit MAb DFS Item
Manufacturer:  Sino Biological 10046R390P25
This antibody was obtained from a rabbit immunized with purified, recombinant Human SPARCL1 (rh SPARCL1; Catalog#10046-H08H; NP_004675.3; Met 1-Phe 664) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-161-1315