Learn More
Sino Biological IL9R Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100461T0850
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human IL9R, and purified by antigen affinity chromatography.
Catalog No. 50-161-1278