Learn More
Sino Biological IL20 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100463T36100
Produced in rabbits immunized with A synthetic peptide corresponding to the center region of the mouse mIL20 , and purified by antigen affinity chromatography.
Catalog No. 50-161-1279