Learn More
Sino Biological CD37 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100465T3250
Produced in rabbits immunized with A synthetic peptide corresponding to the center region of the human CD37, and purified by antigen affinity chromatography.
Catalog No. 50-161-1287