Learn More
Sino Biological Human TrkB / NTRK2 Protein (His & Fc Tag) DFS Item
Manufacturer:  Sino Biological 10047H03H200
A DNA sequence encoding the extracellular domain (Met 1-His 430) of human TrkB (NP_001007098.1) precursor was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the carboxy-terminus.
Catalog No. 50-161-3872