Learn More
Sino Biological TrkB / NTRK2 Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10047MM011
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human TrkB / NTRK2 (rh TrkB / NTRK2; Catalog#10047-H08H; NP_001007098.1; Met1-His430). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-161-1334