Learn More
Sino Biological TrkB / NTRK2 Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10047MM1250
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human NTRK2 / Trk-B extracellular domain (rh Trk-B; Catalog#10047-H08H; NP_001007098.1; Met 1-His 430). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography .
Catalog No. 50-161-1340