Learn More
Sino Biological TrkC / NTRK3 Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10048MM06100
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Trk-C / NTRK3 (rh Trk-C; Catalog#10048-H08H; NP_001007157.1; Met 1-Asp 427). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-161-1372