Learn More
Sino Biological TrkC / NTRK3 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10048RP02100
Produced in rabbits immunized with purified, recombinant Human Trk-C (rhTrk-C; Catalog#10048-H08H; NP_001007157.1; Met 1-Asp 427). Trk-C specific IgG was purified by human Trk-C affinity chromatography.
Catalog No. 50-161-1390