Learn More
Sino Biological LILRA5 / CD85f Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100481T32200
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human LILRA5/CD85f, and purified by antigen affinity chromatography.
Catalog No. 50-161-1350