Learn More
Sino Biological Human CASP7 / caspase 7 / MCH3 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10049H08E10
A DNA sequence encoding the human CASP7 (P55210-1) (Met 1-Gln 303) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-3879