Learn More
Sino Biological KIR3DL3 / CD158z Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100494T4050
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the human KIR3DL3 / CD158z, and purified by antigen affinity chromatography.
Catalog No. 50-161-1398