Learn More
Sino Biological Human CASP3 / Caspase-3 / CASP-3 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10050H08E5
A DNA sequence encoding the mature form of human caspase3 (Q5IS54) (Ser29-His277) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-3882