Learn More
Sino Biological SALL4 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100506T0250
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human SALL4, and purified by antigen affinity chromatography.
Catalog No. 50-161-1419