Learn More
Sino Biological CD13 / ANPEP Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10051MM01200
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD13 / ANPEP (rh CD13 / ANPEP; Catalog#10051-H08H; NP_001141.2; Lys69-Lys967,603Met/Ile). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-161-1441