Learn More
Sino Biological CD13 / ANPEP Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10051RP02100
Produced in rabbits immunized with purified, recombinant Human ANPEP (rh ANPEP; Catalog#10051-H08H; NP_001141.2; Lys 69-Lys 967). ANPEP specific IgG was purified by human ANPEP affinity chromatography .
Catalog No. 50-161-1449