Learn More
Sino Biological IL12B / NKSF2 / P40 Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10052MM01200
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL12B (rhIL12B; Catalog#10052-H08H; Met 1-Ser 328; NP_002178.2). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-161-1473