Learn More
Sino Biological IL12B / IL-12B Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10052MM05100
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL12B / IL-12B (rh IL12B / IL-12B; Catalog#10052-H08H; NP_002178.2; Met1-Ser328). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-161-1476