Learn More
Sino Biological MYL3 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100528T32200
Produced in rabbits immunized with A synthetic peptide corresponding to the N-terminus of the human MYL3, and purified by antigen affinity chromatography.
Catalog No. 50-161-1464