Learn More
Sino Biological NPM1 / Nucleophosmin Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10053T16200
Produced in rabbits immunized with purified, recombinant Human NPM1 / Nucleophosmin (rh NPM1 / Nucleophosmin; Catalog#10053-H07E; P06748-1; Met9-Leu158). NPM1 / Nucleophosmin specific IgG was purified by Human NPM1 / Nucleophosmin affinity chromatography.
Catalog No. 50-161-1501