Learn More
Sino Biological Human BLyS / TNFSF13B / BAFF Protein DFS Item
Manufacturer:  Sino Biological 10056HNCH1
A DNA sequence encoding the soluble form of human BAFF (Q9Y275-1) (Ala 134-Leu 285) was expressed and purified.
Catalog No. 50-161-3902