Learn More
Sino Biological Human I-309 / CCL1 / TCA-3 Protein DFS Item
Manufacturer:  Sino Biological 10057HNAE10
A DNA sequence encoding the human CCL1 (P22362) (Lys24-Lys96) was expressed, with a N-terminal Met.
Catalog No. 50-161-3909