Learn More
Sino Biological NF1B Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100574T3250
Produced in rabbits immunized with A synthetic peptide corresponding to the C-terminus of the human NF1B, and purified by antigen affinity chromatography.
Catalog No. 50-161-1577