Learn More
Sino Biological Numb Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100576T3650
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Numb, and purified by antigen affinity chromatography.
Catalog No. 50-161-1583