Learn More
Sino Biological Human MAPT / Tau Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10058H07E20
A DNA sequence encoding the mature form of human MAPT (NP_058525.1) (Ala2-Leu352) was expressed with a polyhistide tag at the N-terminus.
Catalog No. 50-161-3911