Learn More
Sino Biological HLA-DR Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100586T08200
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human HLA-DR, and purified by antigen affinity chromatography.
Catalog No. 50-161-1597