Learn More
Sino Biological Musashi-1 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100587T10200
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human Musashi-1, and purified by antigen affinity chromatography.
Catalog No. 50-161-1603