Learn More
Sino Biological DPP6 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100592T3250
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human DPP6, and purified by antigen affinity chromatography.
Catalog No. 50-161-1619