Learn More
Sino Biological Human CD157 / BST1 Protein (His Tag) 100ug
Supplier: Sino Biological 10060H08H100
A DNA sequence encoding the human BST1 (NP_004325.2) (Met 1-Lys 292) with a C-terminal polyhistidine tag was expressed.