Learn More
Sino Biological CD157 / BST1 Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10060MM0550
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human BST-1 / CD157 (rh BST-1; Catalog#10060-H08H; Met 1-Lys 292; NP_004325.2). Total IgG was purified by Protein A affinity chromatography .
Catalog No. 50-161-1643