Learn More
Sino Biological CD157/BST1 Antibody (APC), Mouse MAb DFS Item
Manufacturer:  Sino Biological 10060MM10A100
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD157/BST1 (rh CD157/BST1; Catalog#10060-H08H; NP_004325.2; Met1-Lys292) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-161-1647