Learn More
Sino Biological CD157 / BST1 Antibody, Rabbit MAb DFS Item
Manufacturer:  Sino Biological 10060R029100
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD157 / BST1 (rh CD157 / BST1; Catalog#10060-H08H; Met 1-Lys 292; NP_004325.2).
Catalog No. 50-161-1662