Learn More
Sino Biological Oct4 / POU5F1 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100616T0850
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Mouse Oct4 / POU5F1, and purified by antigen affinity chromatography.
Catalog No. 50-161-1684