Learn More
Sino Biological BACE1 / ASP2 Antibody, Rabbit MAb DFS Item
Manufacturer:  Sino Biological 10064R001100
This antibody was obtained from a rabbit immunized with purified, recombinant Human BACE1 extracellular domain (rh BACE1; Catalog#10064-HCCH; Met 1-Thr 457; NP_036236.1) .
Catalog No. 50-161-1726