Learn More
Sino Biological BACE1 Antibody, Rabbit MAb DFS Item
Manufacturer:  Sino Biological 10064R027100
This antibody was obtained from a rabbit immunized with purified, recombinant Human BACE1 (rh BACE1; Catalog#10064-H08H; NP_036236.1; Met1-Thr457).
Catalog No. 50-161-1728