Learn More
Sino Biological BACE1 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10064T16100
Produced in rabbits immunized with purified, recombinant Human BACE1 (rh BACE1; Catalog#10064-H08H; NP_036236.1; Met1-Thr457). BACE1 specific IgG was purified by Human BACE1 affinity chromatography.
Catalog No. 50-161-1737